missing translation for 'onlineSavingsMsg'
Learn More

Starch, Soluble, ACS Reagent, Reagents

Catalog No. 01337654
Change view
Click to view available options
Quantity:
125 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337654 125 g
01337652 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337654 Supplier Labchem Inc Supplier No. C1332000125D2
Only null left

Starch, Soluble, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 9005-84-9
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
InChI Key GUBGYTABKSRVRQ-ASMJPISFSA-N
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
SMILES OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Boiling Range 668°C
Color White
Boiling Point 668°C
Packaging Glass Bottle
Quantity 125 g
Specific Gravity 1.5
Grade ACS
Physical Form Solid
Chemical Name or Material Starch, Soluble, ACS Reagent