missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch Soluble ACS, MP Biomedicals™
$84.01 - $84.01
Chemical Identifiers
| CAS | 9005-84-9 |
|---|---|
| Molecular Formula | C12H22O11 |
| Molecular Weight (g/mol) | 342.297 |
| MDL Number | MFCD00082026 |
| InChI Key | GUBGYTABKSRVRQ-ASMJPISFSA-N |
| Synonym | alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose |
| PubChem CID | 439341 |
| ChEBI | CHEBI:18167 |
| IUPAC Name | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol |
| SMILES | C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15258280
|
MP Biomedicals Inc
0215258280 |
100 g |
Each for $84.01
|
|
|||||
Chemical Identifiers
| 9005-84-9 | |
| 342.297 | |
| GUBGYTABKSRVRQ-ASMJPISFSA-N | |
| 439341 | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol |
| C12H22O11 | |
| MFCD00082026 | |
| alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
| CHEBI:18167 | |
| C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O |
Specifications
| 9005-84-9 | |
| C12H22O11 | |
| 100 g | |
| GUBGYTABKSRVRQ-ASMJPISFSA-N | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol | |
| 439341 | |
| 342.3 | |
| Powder |
| White | |
| MFCD00082026 | |
| alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
| C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O | |
| 342.297 | |
| CHEBI:18167 | |
| ACS | |
| Starch Soluble |
Safety and Handling
Recommended Storage : Store at Room Temperature (15-30°C).