missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch, MP Biomedicals
Specifications
Specifications
| Quantity | 100 lb. |
| Physical Form | Fine Powder |
| Chemical Name or Material | Starch, From Corn |
Safety and Handling
| Recommended Storage | Room Temperature |