missing translation for 'onlineSavingsMsg'
Learn More

Starch indicator solution 1%, w/v aqueous solution (for iodometric titrations)

Catalog No. p-7052331
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA47209AP 500 mL
AA47209K2 1 L
AA47209K7 4 L
AA47209M4 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA47209AP Supplier Thermo Scientific Chemicals Supplier No. 047209AP
Only null left

For iodometric titrations | CAS: 9005-84-9 | C12H22O11 | 342.297 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9005-84-9
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.297
MDL Number MFCD00082026
InChI Key GUBGYTABKSRVRQ-ASMJPISFSA-N
Synonym alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
PubChem CID 439341
ChEBI CHEBI:18167
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O

Specifications

Physical Form Liquid for iodometric titrations
Merck Index 14,8799
Quantity 500 mL
Chemical Name or Material Starch indicator solution 1%, w/v aqueous solution for iodometric titrations
EINECSNumber 232-686-4
RTECSNumber GM5090000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only