missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch Indicator, Preserved with Salicylic Acid (Mercury Free), Certified, 1%, LabChem™
Supplier: LabChem LC253102
Specifications
| Starch Indicator | |
| 98.9,1,0.1 | |
| 10% | |
| Poly Bottle | |
| Liquid |
| Preserved with Salicylic Acid (Mercury Free) | |
| 9005-25-8,69-72-7 | |
| Certified | |
| 1 L |