missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch Indicator, Preserved with Salicylic Acid (Mercury Free), Certified, 1%, LabChem™
Supplier: LabChem LC253101
Specifications
| Starch Indicator | |
| 98.9,1,0.1 | |
| Soluble in water | |
| 1% | |
| Poly Bottle | |
| Passes Test | |
| 1g/mL | |
| 500 mL |
| Preserved with Salicylic Acid (Mercury Free) | |
| 9005-25-8,69-72-7 | |
| Passes Test | |
| Certified | |
| 1g/mL | |
| Fume; Carbon monoxide; Carbon dioxide | |
| Colorless to Pink | |
| Liquid |