missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch Indicator, Preserved with Salicylic Acid (Mercury Free), Certified, 0.5%, LabChem™
$52.93 - $732.75
Specifications
| Chemical Name or Material | Starch Indicator |
|---|---|
| Name Note | Preserved with Salicylic Acid (Mercury Free) |
| CAS Max % | 99.4,0.5,0.1 |
| CAS | 9005-25-8 , 69-72-7 |
| Solubility Information | Soluble in water |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
LC253001
|
LabChem
LC253001 |
500 mL | Poly Bottle |
Each for $52.93
|
|
||||
|
LC253002
|
LabChem
LC253002 |
1 L | Poly Bottle |
Each for $77.50
|
|
||||
|
LC253004
|
LabChem
LC253004 |
4 L | Poly Bottle |
Each for $197.04
|
|
||||
|
LC253005
|
LabChem
LC253005 |
20 L | Cubitainer™ |
Each for $732.75
|
|
||||