missing translation for 'onlineSavingsMsg'
Learn More

Starch Indicator, Preserved with Salicylic Acid (Mercury Free), Certified, 0.2%, LabChem™

Supplier:  LabChem LC252907

Catalog No. LC252907

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Starch Indicator
99.7,0.2,0.1
Soluble in water
0.2%
Poly Bottle
Passes Test
1g/mL
Liquid
Preserved with Salicylic Acid (Mercury Free)
9005-25-8 , 69-72-7
Passes Test
Certified
1g/mL
Fume; Carbon monoxide; Carbon dioxide
125 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions