missing translation for 'onlineSavingsMsg'
Learn More

Starch Indicator, 1% (w/v), Mercury Free, for Iodometric Titrations, Ricca Chemical

Catalog No. 805016
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
805016 500 mL Natural Poly Bottle
805032 1 L Natural Poly Bottle
R80500004F 4 L Cubitainer
80501 4 L Natural Poly Bottle
805025 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 805016 Supplier Ricca Chemical Company Supplier No. 805016
Edge

Iodometric Titrations

Specifications

Chemical Name or Material Starch Indicator
CAS 7732-18-5, 9005-84-9, 69-72-7
CAS Min % 97.0, 0.97, 0.12
CAS Max % 100.0, 1.01, 0.13
Concentration 1% (w/v)
Percent Purity 1% W/V
Grade Indicator
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color White
pH 5 to 7
Quantity 500 mL
Physical Form Liquid
Show More Show Less