missing translation for 'onlineSavingsMsg'
Learn More

Starch Indicator, 0.5% (w/v), Mercury Free, for Iodometric Titrations, Ricca Chemical

Catalog No. R8000000100
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R8000000100 100 mL Natural Poly Bottle
800016 500 mL Natural Poly Bottle
800032 1 L Natural Poly Bottle
80001 4 L Natural Poly Bottle
80005 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. R8000000100 Supplier Ricca Chemical Company Supplier No. R8000000100A

Iodometric Titrations

Specifications

Chemical Name or Material Starch Indicator
CAS 9005-84-9, 69-72-7, 7732-18-5
Concentration 0.5% (w/v)
Percent Purity 0.5% W/V
Grade Indicator
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color White
pH 5 to 7
Quantity 100 mL
Physical Form Liquid
Show More Show Less