missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch, Extracted from Potato, MP Biomedicals™
Starch is used as an additive for food processing, food starches are typically used as thickeners and stabilizers
$315.00 - $315.00
Chemical Identifiers
| CAS | 9005-25-8 |
|---|---|
| Molecular Formula | C12H22O11 |
| Molecular Weight (g/mol) | 342.30 |
| MDL Number | MFCD00082026,MFCD00132834 |
| InChI Key | GUBGYTABKSRVRQ-UHFFFAOYNA-N |
| Synonym | alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose |
| PubChem CID | 439341 |
| ChEBI | CHEBI:18167 |
| IUPAC Name | 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol |
| SMILES | OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10295402
|
MP Biomedicals Inc
0210295402 |
2 kg |
Each for $315.00
|
|
|||||
Chemical Identifiers
| 9005-25-8 | |
| 342.30 | |
| GUBGYTABKSRVRQ-UHFFFAOYNA-N | |
| 439341 | |
| 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol |
| C12H22O11 | |
| MFCD00082026,MFCD00132834 | |
| alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
| CHEBI:18167 | |
| OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O |
Specifications
| Powder | |
| C12H22O11 | |
| alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
| GUBGYTABKSRVRQ-UHFFFAOYNA-N | |
| 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol | |
| 439341 | |
| Starch |
| 9005-25-8 | |
| MFCD00082026,MFCD00132834 | |
| 2 kg | |
| OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O | |
| 342.30 | |
| CHEBI:18167 |
Safety and Handling
EINECSNumber : 232-679-6