missing translation for 'onlineSavingsMsg'
Learn More

Starch from corn for use with Total Dietary Fiber Control Kit, TDF-C10, MilliporeSigma™ Supelco™

Catalog No. 111015827
Change view
Click to view available options
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111015827 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015827 Supplier Merck Emd Millipore Supplier No. S238810G
Only null left
Food and Beverages and General Analytical

Specifications

CAS 9005-25-8
Quantity 10 g
Linear Formula (C6H10O5)n
MDL Number MFCD00082026
Synonym Corn starch; Starch from maize
Packaging Poly Bottle