missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Starch, for biochemistry, potato, hydrolyzed for electrophoresis

Catalog No. p-29623
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC215121000 100 g Plastic bottle
AC215125000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC215121000 Supplier Thermo Scientific Chemicals Supplier No. 215121000
Only null left

CAS: 9005-25-8 | C12H22O11 | 342.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9005-25-8
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00082026,MFCD00132834
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
Synonym alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
PubChem CID 439341
ChEBI CHEBI:18167
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Specifications

Color White
Physical Form Fine Crystalline Powder
Merck Index 14, 8798
Solubility Information Solubility in water: insoluble.
Quantity 100 g
Grade Biochemical
Infrared Spectrum Authentic
Loss on Drying 20% max.
Packaging Plastic bottle
Residue after Ignition 1% max.
Chemical Name or Material Starch, Hydrolyzed for Electrophoresis
Show More Show Less

RUO – Research Use Only