missing translation for 'onlineSavingsMsg'
Learn More

Starch, Arrowroot, Powder, Spectrum™ Chemical

Catalog No. 18606999
Change view
Click to view available options
Quantity:
2.5 kg
6 x 500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606999 2.5 kg Poly Bottle
18606998 6 x 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18606999 Supplier Spectrum Chemical Mfg Cor Supplier No. S15512.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions

S1551, 9005-25-8, (C6H10O5)n

Spectrum™ Chemical Starch, Arrowroot, Powder is used in talcum powders and hair dyes. It is added to moisturizers as a thickening agent and to help active ingredients penetrate the upper levels of the skin. This product is used in cosmetics to help moisturizers penetrate the skin. It appears as a fine, white powder. It may have other uses similar to cornstarch. In cosmetics, it is used as an abrasive.

Chemical Identifiers

CAS 9005-25-8
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00082026,MFCD00132834
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
IUPAC Name 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Specifications

CAS Max % 1
Quantity 2.5 kg
Grade Reagent
Packaging Poly Bottle