missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch, ACS reagent, soluble, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 424490100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Starch | |
| 9005-84-9 | |
| MFCD00082026 | |
| 10kg | |
| C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O | |
| 342.297 | |
| CHEBI:18167 | |
| Authentic | |
| 0.4% max. |
| Soluble | |
| C12H22O11 | |
| alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
| GUBGYTABKSRVRQ-ASMJPISFSA-N | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol | |
| 439341 | |
| ACS Reagent | |
| Plastic drum | |
| Passes Test |
Safety and Handling
EINECSNumber : 232-686-4
TSCA : TSCA
RUO â Research Use Only