missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Staphylase™ Test

Catalog No. OXDR0595A
Change view
Click to view available options
Quantity:
100 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXDR0595A 100 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXDR0595A Supplier Thermo Scientific™ Supplier No. DR0595A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rapidly identify Staphylococcus aureus with this slide agglutination test which detects the presence of the clumping factor from selective or nonselective culture media.

The generally accepted identifying characteristic of Staphylococcus aureus is the ability to produce free and bound coagulase (or clumping factor). The presence of clumping factor may be detected in a number of ways. The Oxoid Staphylase Test detects the presence of clumping factor through clumping of fibrinogen- sensitized sheep red blood cells1,2. The specificity of the reaction is ensured by a simultaneous test with a control reagent (unsensitized sheep red blood cells), when of course no clumping reaction should be observed.

Use this rapid agglutination test for the identification of Staphylococcus aureus by detection of clumping factor. Clumping factor is detected through agglutination of fibrinogen-sensitized sheep red blood cells. The specificity of the reaction is ensured by a simultaneous test with a control reagent (unsensitized sheep red blood cells) when no clumping reaction should be observed. A positive result is obtained if clumping of the test cell suspensions occurs during mixing. This indicates the presence of Staphylococcus aureus.

Included in the kit is a test reagent consisting of rabbit fibrogen-sensitized sheep red blood cells, control reagent consisting of unsensitized sheep red blood cells, and disposable reaction cards.

Each kit contains sufficient reagent for 100 tests.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications

Content And Storage 2°C to 8°C
Control Sets Positive
Description Staphylase Test
Detectable Analytes S. aureus
Type Staphylococcus aureus Test Kit
Includes Test reagent, control reagent, disposable reaction cards. Each kit contains sufficient reagent for 100 tests.
Quantity 100 Tests/Kit
Target Organism Class Staphylococcus aureus