missing translation for 'onlineSavingsMsg'
Learn More

MAPA™ StanSolv™ A-14 Unlined Nitrile Gloves

Catalog No. 113924C
Change view
Click to view available options
Size:
8
9
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
113924C 9
113924B 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 113924C Supplier MAPA™ Supplier No. 474019

Heavy-weight, unlined chemical protection nitrile gloves feature a z-pattern grip for a secure handling of hard-to-grasp items.

  • Z-Patter grip channels liquids away from glove surface for an enhanced grip in wet applications
  • Nitrile compound provides resistance to snags, punctures, abrasions and cuts
  • Chlorinated finish provides easy on/off
  • Straight cuff

Specifications

Certifications/Compliance 21 CFR 170-199
Certifications/Compliance (Europe) Cat III CE 0334, EN 374
Chlorinated Yes
Color Green
Latex Free Yes
Material Nitrile
Size 9
Thickness (English) 22 mil
Thickness (Metric) 0.55 mm
EN388 Standard EN 388:4102
EN 421 No
EN ISO 374-1 CHEMICAL LETTER CODES AJKL
EN ISO 374 - 5 Bacteria, Fungi only
For Use With (Application) Handling drums of chemical products, etching/degreasing, treating/degreasing metals with aggressive solvents, pesticide application, maintenance, timber treatment and finishing
Glove Cuff Style Straight
Grip Design Z-pattern
Lining Unlined
Model 474019
Series StanSolv™ A-14
Length (English) 14 in.
Length (Metric) 35.5 cm
Show More Show Less