missing translation for 'onlineSavingsMsg'
Learn More

Stannous Sulfate, Technical, Spectrum™ Chemical

$730.63 - $15366.47

Chemical Identifiers

CAS 7488-55-3
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18614052
View Documents
Spectrum Chemical Mfg Cor
S1550500GM
500 g Poly Bottle
Each for $730.63
Only null left
 
18614050
View Documents
Spectrum Chemical Mfg Cor
S155025KG
2.5 kg Poly Pail
Each for $2,773.63
Only null left
 
18614051
View Documents
Spectrum Chemical Mfg Cor
S155025KGBL
25 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7488-55-3

Specifications

7488-55-3
500 g
Poly Bottle
1
Technical
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.