missing translation for 'onlineSavingsMsg'
Learn More

Stannous Chloride, Dihydrate, Technical, 98.0-103.0%, Spectrum™ Chemical

Catalog No. 18614033
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614033 50 kg Poly Bottle
18614030 500 g Poly Bottle
18614031 2.5 kg Poly Bottle
18614032 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18614033 Supplier Spectrum Chemical Mfg Cor Supplier No. S153650KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10025-69-1

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 50 kg
Molecular Weight (g/mol) 225.65
Percent Purity 98 to 103%
Grade Technical
Packaging Poly Bottle
Chemical Name or Material Stannous Chloride, Dihydrate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.