missing translation for 'onlineSavingsMsg'
Learn More

Stannous Chloride Dihydrate, Purified (Tin (II) Chloride Dihydrate), Reagents

Catalog No. 01336947
Change view
Click to view available options
Quantity:
100 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336947 100 g
01336946 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336947 Supplier Labchem Inc Supplier No. C2330000100C2
Only null left

Stannous Chloride Dihydrate, Purified (Tin (II) Chloride Dihydrate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10025-69-1
Molecular Formula Cl2H4O2Sn
Molecular Weight (g/mol) 225.64
InChI Key FWPIDFUJEMBDLS-UHFFFAOYSA-L
Synonym Tin(II) Chloride Dihydrate
IUPAC Name λ²-tin(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Sn++]

Specifications

Boiling Point 652°C
Color White to Off-White
Physical Form Solid
Quantity 100 g
Grade Pure
Packaging Glass Bottle
Specific Gravity 2.71
Chemical Name or Material Stannous Chloride Dihydrate, Purified (Tin (II) Chloride Dihydrate)