missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorure d’étain(II) dihydraté (cristallin, certifié ACS), Fisher Chemical™
Spécifications
Spécifications
| Point d’ébullition | 652 °C |
| Nom ou substance chimique | Stannous Chloride Dihydrate |
| Point de fusion | 37 °C |
| Couleur | Incolore |
| Forme physique | Solide |
| Quantité | 500 g |
| Remarques sur le degré de pureté | Adapté à la détermination du mercure |
| Fourchette de pourcentages d’essai | 98 à 103% |
| Renseignements sur la solubilité | Soluble dans l’eau |
| Poids de formule | 225.65 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Causes burns by all exposure routes. May cause an allergic skin reaction. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting. NFPA Health:3 Flammability:0 Instability:1 |
| DOTInformation | DOT Class 8, : Corrosive |