missing translation for 'onlineSavingsMsg'
Learn More

Stannous Chloride Dihydrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. T142100
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
T142100 100 g Poly Bottle
T142500 500 g Poly Bottle
T1423 3 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T142100 Supplier Thermo Fisher Scientific Supplier No. T142100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10025-69-1
Molecular Formula Cl2Sn · 2 H2O
MDL Number MFCD00149863
Synonym Tin (II) Chloride Dihydrate

Specifications

Boiling Point 652°C
Melting Point 37°C
Color White
Physical Form Solid
Quantity 100 g
Assay Percent Range 98 to 103 %
Solubility Information Soluble in water
Formula Weight 225.65
Percent Purity 98 to 103%
Grade Suitable for mercury determination
Identification Pass test
Packaging Poly Bottle
Chemical Name or Material Stannous Chloride Dihydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Causes burns by all exposure routes. May cause an allergic skin reaction. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:0
Instability:1
DOTInformation DOT Class 8, : Corrosive