missing translation for 'onlineSavingsMsg'
Learn More

Stannous Chloride Dihydrate, ACS Reagent Grade, Ricca Chemical

Catalog No. rdcs1010500
Change view
Click to view available options
Quantity:
500 g
Packaging:
White Poly Wide Mouth Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCS1010500 500 g White Poly Wide Mouth Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCS1010500 Supplier Ricca Chemical Company Supplier No. RDCS1010500B1

Chemical Identifiers

CAS 10025-69-1
Molecular Weight (g/mol) 225.65 g/mol

Specifications

Color Colorless
CAS Min % 98.0
CAS Max % 100.0
Physical Form Crystalline
Quantity 500 g
Percent Purity 98 to 103%
Grade ACS
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Stannous Chloride Dihydrate