missing translation for 'onlineSavingsMsg'
Learn More

Stannous Chloride, Dihydrate, ACS Grade, 98.0% to 103.0%, LabChem™

Catalog No. lc251701
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC251701 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC251701 Supplier LabChem Supplier No. LC251701
Only null left

Chemical Identifiers

CAS 10025-69-1
Synonym Tin Chloride

Specifications

Melting Point 38°C
Color White
CAS Max % 100
Physical Form Crystals
Quantity 500 g
Linear Formula SnCl2·2H2O
UN Number UN3260
Solubility Information Soluble in water
Formula Weight 225.65
Percent Purity 98.0 to 103.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 2710kg/m3
Decomposition Information Hydrogen chloride
Density 2,710kg/m3
Chemical Name or Material Stannous Chloride, Dihydrate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Do not breathe dust.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature