missing translation for 'onlineSavingsMsg'
Learn More

Stannous Chloride, Dihydrate, 98.0 to 101.0%, ACS, MilliporeSigma™

Catalog No. MSX08855
Change view
Click to view available options
Quantity:
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSX08855 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSX08855 Supplier Technidata Supplier No. 1.07815.1000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10025-69-1
Molecular Formula SnCl2·2H2O
Synonym Tin(II) Chloride, Dihydrate

Specifications

Color White
Physical Form Crystalline Solid
Quantity 2.5 kg
Assay Percent Range 98.0 to 101.0%
Formula Weight 225.65
Chemical Name or Material Stannous Chloride, Dihydrate, For ACS analysis