missing translation for 'onlineSavingsMsg'
Learn More

Stannous Chloride, Anhydrous, 96%, Spectrum™ Chemical

Catalog No. p-7166382
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
12 kg
2.5 kg
25 g
45 kg
500 g
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18614026 25 g Amber Glass Bottle
18614027 100 g Poly Bottle
18614029 500 g Poly Bottle
18614024 2.5 kg Poly Bottle
18614028 12 kg Poly Pail
18614025 45 kg Fiber Drum
6 options

Catalog No. 18614026

Supplier: Spectrum Chemical Mfg Cor S153325GM

Only null left

Chemical Identifiers

CAS 7772-99-8

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 96%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.