missing translation for 'onlineSavingsMsg'
Learn More

Stannous Chloride, Anhydrous, 96%, Spectrum™ Chemical

Catalog No. 18614026
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614026 25 g Amber Glass Bottle
18614027 100 g Poly Bottle
18614029 500 g Poly Bottle
18614024 2.5 kg Poly Bottle
18614028 12 kg Poly Pail
18614025 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18614026 Supplier Spectrum Chemical Mfg Cor Supplier No. S153325GM
Only null left

Chemical Identifiers

CAS 7772-99-8

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 96%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.