missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Standing Height Class 100 Clean Room Vinyl Sit-Stand with Stationary Glides

Catalog No. 15004007
Change view
Click to view available options
Color:
Adobe White
Black
Bluebird
Borscht
Burgundy
Carbon
Charcoal
Colonial Blue
Dove
Elephant Gray
Graphite
Imperial Blue
Ink
Mandarin
Marine Blue
Orange Kist
Sterling
Storm
Taupe
Material:
Vinyl (Grade A)
Vinyl (Grade B)
Vinyl (Grade C)
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material
15004007 Orange Kist Vinyl (Grade B)
15002466 Mandarin Vinyl (Grade C)
15002542 Imperial Blue Vinyl (Grade B)
15002595 Bluebird Vinyl (Grade C)
15002861 Black Vinyl (Grade B)
15002862 Ink Vinyl (Grade C)
15002863 Colonial Blue Vinyl (Grade B)
15002864 Taupe Vinyl (Grade A)
15003115 Marine Blue Vinyl (Grade A)
15003116 Taupe Vinyl (Grade B)
15003117 Sterling Vinyl (Grade A)
15003192 Burgundy Vinyl (Grade B)
15003456 Borscht Vinyl (Grade A)
15003488 Adobe White Vinyl (Grade B)
15003489 Elephant Gray Vinyl (Grade C)
15003519 Charcoal Vinyl (Grade B)
15003896 Black Vinyl (Grade C)
15003922 Storm Vinyl (Grade A)
15003924 Carbon Vinyl (Grade A)
15003993 Graphite Vinyl (Grade C)
15003994 Dove Vinyl (Grade B)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Catalog No. 15004007 Supplier Fisherbrand™ Supplier No. 91CLRV970338613
Get 1 Extra Fisherbrand Chair When You Buy 4
View all promotions

Vinyl standing height sit-stand.

  • Adjustable from 21 to 31 in. from the top of the seat to the floor
  • Features seat height adjustment
  • 8-degree forward and 3-degree backward locking seat tilt
  • Cast-aluminum base
  • Black non-marring bell glides (suitable for both hard and carpeted floors)
  • Variety of color options to suit any work space
  • Class 100/ISO 5 Clean Room

Specifications

Product Type Controlled Environment Chair
Footrest No
With Casters No
Color Orange Kist
With Backrest No
With Armrest No
Autoclavable Not Autoclavable
With Lumbar Support No
With Tilt Yes
Material Vinyl (Grade B)
Seating Type High Bench Height
Show More Show Less