missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Standing Height Class 100 Clean Room Vinyl Sit-Stand with Chrome Foot Ring and Stationary Glides

Catalog No. 15003995
Change view
Click to view available options
Color:
Adobe White
Black
Bluebird
Borscht
Burgundy
Carbon
Charcoal
Colonial Blue
Dove
Elephant Gray
Graphite
Imperial Blue
Ink
Mandarin
Marine Blue
Orange Kist
Sterling
Storm
Taupe
Material:
Vinyl (Grade A)
Vinyl (Grade B)
Vinyl (Grade C)
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material
15003995 Adobe White Vinyl (Grade B)
15002433 Black Vinyl (Grade B)
15002434 Dove Vinyl (Grade B)
15002486 Taupe Vinyl (Grade B)
15002543 Bluebird Vinyl (Grade C)
15002544 Elephant Gray Vinyl (Grade C)
15002596 Black Vinyl (Grade C)
15002839 Imperial Blue Vinyl (Grade B)
15002840 Taupe Vinyl (Grade A)
15003099 Storm Vinyl (Grade A)
15003158 Orange Kist Vinyl (Grade B)
15003438 Burgundy Vinyl (Grade B)
15003439 Marine Blue Vinyl (Grade A)
15003440 Sterling Vinyl (Grade A)
15003448 Carbon Vinyl (Grade A)
15003449 Borscht Vinyl (Grade A)
15003490 Mandarin Vinyl (Grade C)
15003518 Ink Vinyl (Grade C)
15003849 Colonial Blue Vinyl (Grade B)
15003850 Charcoal Vinyl (Grade B)
15003923 Graphite Vinyl (Grade C)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Catalog No. 15003995 Supplier Fisherbrand™ Supplier No. 91CLRV97033CF8501
Get 1 Extra Fisherbrand Chair When You Buy 4
View all promotions

Vinyl standing height sit-stand with chrome foot ring and stationary glides.

  • Adjustable from 21 to 31 in. from the top of the seat to the floor
  • Features seat height adjustment
  • 18 in. diameter round tube chrome foot ring with knob lock
  • 8-degree forward and 3-degree backward locking seat tilt
  • Cast-aluminum base
  • Black non-marring bell glides (suitable for both hard and carpeted floors)
  • Variety of color options to suit any work space
  • Class 100/ISO 5 Clean Room

Specifications

Product Type Controlled Environment Chair
With Tilt Yes
Footrest Yes
With Lumbar Support No
With Casters No
With Backrest No
With Armrest No
Color Adobe White
Autoclavable Not Autoclavable
Material Vinyl (Grade B)
Seating Type High Bench Height
Show More Show Less