missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Standard Vehicle Placards: NON-FLAMMABLE GAS 2
Indicate delivery vehicles carrying non flammable gas materials. Brady™ Standard Vehicle Placards are available in a variety of materials.
$4.38
Specifications
| Adhesive Type | Non-adhesive |
|---|---|
| Color | White, Green |
| Display Text | NON-FLAMMABLE GAS 2 |
| For Use With (Application) | Material Handling, Shipping and Receiving Signs and Labels |
| Quantity | 1 Ea. |
Description
- Available in two different materials Polycoated Tagstock and Premium Fiberglass
Specifications
| Non-adhesive | |
| NON-FLAMMABLE GAS 2 | |
| 1 Ea. | |
| 10.75 x 10.75 in. |
| White, Green | |
| Material Handling, Shipping and Receiving Signs and Labels | |
| Diamond |