Learn More
Beckman Coulter Standard Vacuum Unit
Catalog No. 15655168
Change view
Click to view available options
Product Type:
Standard Vacuum Unit
Catalog No. Product Type
15655168 Standard Vacuum Unit
1 options
Catalog No. 15655168 Supplier Beckman Coulter Supplier No. 7809200
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Designed to be used with the Dry Powder System (DPS) of the LS 13 320 XR or the Tornado Module of the LS 13 320 for delivering dry powder samples to the sensing zone in the optical bench of the Particle Size Analyzer.
  • A vacuum unit is required for use with the Dry Powder System (DPS) Module and the LS 13 320 XR Particle Size Analyzer
  • Standard Vacuum Unit features a fully adjustable pressure range
  • Replacement vacuum bags available
Specifications
Product Type Standard Vacuum Unit
For Use With (Application) Delivering Dry Powder Samples to the Sensing Zone in the Optical Bench
For Use With (Equipment) LS 13 320 XR, or LS 13 320 Particle Size Analyzer with Dry Powder System (DPS)