Learn More
Brady™ Standard Traffic Signs
Catalog No. 19017679
Change view
Click to view available options
Color:
Black, White
Black, Yellow
White, Red
Dimensions (L x W):
18 x 18 in.
24 x 18 in.
30 x 30 in.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Dimensions (L x W)
19017679 White, Red 18 x 18 in.
19041960 Black, Yellow 30 x 30 in.
19017681 White, Red 30 x 30 in.
19017640 Black, White 24 x 18 in.
19017686 Black, White 24 x 18 in.
19017639 Black, White 18 x 18 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19017679 Supplier Brady™ Supplier No. 94142
Meet MUTCD specifications
  • Designed to meet Manual Uniform Traffic Control Devices (MUTCD) specifications
  • Made with B-959 reflective sheeting on aluminum
Specifications
Dimensions (L x W) 18 x 18 in.
Color White, Red