missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bal Supply Standard Taper Joint Clips
Description
- Nickel Plated Steel
- Can withstand temperatures up to 932°F
- For Standard Taper Joints
Specifications
Specifications
| Quantity | 10/Pk. |
| Type | Standard Taper Clamp |
| Material | Stainless Steel |
| For Use With (Equipment) | Standard Taper Joints |
| Size | 14 |
| Color | Silver |