Learn More
Thermo Scientific™ Standard Single-Use Bottle Assembly System, polycarbonate, 3-port cap set, 2 L
GREENER_CHOICE
Catalog No. SB00005I
Change view
Click to view available options
Quantity:
8 Assemblies
Catalog No. Quantity
SB00005I 8 Assemblies
1 options
Catalog No. SB00005I Supplier Thermo Scientific™ Supplier No. SB00005I
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Thermo Scientific™ PETG Single-Use Bottle Assembly System includes a Nalgene Square PETG Bottle, a 38 to 430 mm ported closure with one vent filter line and one 1/8-in plugged connection line.
Key benefits:
  • Designed for a broad range of applications, including bioproduction, life science, and general lab use
  • Helps eliminate setup and post-use cleaning steps required for reuseable containers
  • Helps reduce cross-contamination risks
  • Allows process-specific flexibility with multiple bottle sizes
Assembly details:
  • Bottles are available in polycarbonate (PC) and polyethylene terephthalate glycol (PETG) contact surfaces
  • Each bottle is topped with an assembly including a cap, puck, and tubing
  • Most assemblies contain two ports with C-Flex™ tubing; the 2 L size includes a third port, also with C-Flex tubing
  • One line on each assembly is dedicated to venting
  • The second and third lines are for liquid transfer and include 1/8 in. plug fittings, to be used with your preferred welding or sealing equipment
  • Fluid path of the finished product has been validated according to AAMI TIR33 with a 10-6 sterility assurance level (SAL)
Specifications
Product Type Single Use Mixer Transfer Assembly
Material Polycarbonate
Sterility Sterile
Capacity (English) 67.62 oz.
Capacity (Metric) 2 L
Content And Storage Store at room temperature.
Quantity 8 Assemblies
Closure Material Polypropylene
Closure Type Screw Cap
Through the Closure Dispensing Yes
Workflow Step Upstream sampling. Upstream Validation. Upstream process solution storage.
Show More Show Less