Learn More
BINDER™ Standard Rack
Shelf rack in-set with pair of retaining U-rails, for use with climate chambers and cooling incubator chambers.
Supplier: BINDER™ 80122392
Description
- Shelf rack made of stainless steel
- Includes set of U-rails
Chemical Identifiers
| Stainless Steel | |
| Rack |
Specifications
| KB/KB PRO 1060, KBF/KBF PRO 1060 | |
| Stainless Steel | |
| Rack |