Learn More
BINDER™ Standard Rack
Shelf rack in-set with pair of retaining U-rails, for use with climate chambers and cooling incubator chambers.
Supplier: BINDER™ 80122391
Description
- Shelf rack made of stainless steel
- Includes set of U-rails
Chemical Identifiers
| Stainless Steel | |
| Rack |
Specifications
| KB/KB PRO 720, KBF/KBF PRO 720 | |
| Stainless Steel | |
| Rack |