Learn More
BINDER™ Standard Rack
Description
- Shelf rack made of stainless steel
- Includes set of U-rails
Specifications
Specifications
| For Use With (Equipment) | KB PRO/KBF/KBF PRO 1600 |
| Material | Stainless Steel |
| Type | Rack |