missing translation for 'onlineSavingsMsg'
Learn More

Puritan™ Standard Polyester Swab, Wooden Handle

Catalog No. 22029682
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
22029682 Sterile
361045499 Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22029682 Supplier Puritan™ Supplier No. 25806 1WD
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puritan™ Standard Polyester Swabs are generally used for specimen collection

  • Polyester tip contains no fatty acids or latent sugars
  • Non-absorbent tip
  • Sturdy wood handle is naturally biodegradable

Specifications

Description Polyester tipped applicator with wooden handle.
For Use With (Application) Specimen collection; testing and screening swab; Patient care; Surface collection
Diameter (English) Tip 0.188 in.
Diameter (Metric) Tip 4.78 mm
Length (English) Tip 0.687 in.
Length (English) 6in.
Length (Metric) 150.80 mm
Length (Metric) Tip 17.45 mm
Quantity 1000/Pk., 10 Pk./Cs.
Product Type Polyester Swab
Sterility Sterile
Handle Material Wood
Tip Type Standard
Tip Material Polyester
Show More Show Less