missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Standard Plate Count Agar (APHA) (Dehydrated)
Description
Determine the plate counts of microorganisms in food, dairy samples, waste water and other materials of sanitary importance with this medium.
- Meets formulation of APHA, AOAC and PHLS
- Quality control of Standard Plate Count Agar (APHA) includes testing in accordance with ISO 11133:2014
Specifications
Specifications
| Certifications/Compliance | APHA, AOAC, PHLS, ISO 11133:2014 |
| Form | Powder |
| Product Type | Agar |
| Test Type | Standard Plate Count for Microorganism |
| Quantity | 500 g |
| Yield | For 21.3L medium |
For laboratory use only