missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ Standard PFTE Stopper
With handle for easy gripping.
$88.10 - $215.73
Specifications
| Color | White |
|---|---|
| Material | PTFE |
| For Use With (Application) | Organic Synthesis |
| Quantity | 1 Ea. |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
31501894
|
Synthware™
S111420 |
14/20 |
Each for $88.10
|
|
|||||
|
31501895
|
Synthware™
S111922 |
19/22 |
Each for $91.93
|
|
|||||
|
31501896
|
Synthware™
S112425 |
24/25 |
Each for $118.90
|
|
|||||
|
31501897
|
Synthware™
S112440 |
24/40 |
Each for $122.57
|
|
|||||
|
31501898
|
Synthware™
S112942 |
29/42 |
Each for $174.10
|
|
|||||
|
31501899
|
Synthware™
S113445 |
34/45 |
Each for $215.73
|
|
|||||
Specifications
| White | |
| PTFE | |
| Organic Synthesis | |
| 1 Ea. |