missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Standard Leather Palm Gloves - Clute Cut

Catalog No. 19013463
Change view
Click to view available options
Glove Cuff Style:
Knit
Gender:
Men's
Women's
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Glove Cuff Style Gender
19013463 Knit Men's
19013461 Knit Women's
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19013463 Supplier Fisherbrand™ Supplier No. 650IL

Gloves feature a fabric back for breathability, with split cowhide leather on the palm for increased durability and strength.

  • Fabric back with split cowhide leather on the palm
  • Straight thumb with leather on palm side of the gloves
  • Available with a knitwrist, or a 2.5 in. rubberized safety cuff
  • Clute cut

Specifications

Color Gray, Blue, Red
Grade Standard
Hand Specific
Material Shoulder Split Cowhide Leather
Product Type General Purpose Gloves
Size Large
Style Clute
Gender Men's
Glove Cuff Style Knit
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less