missing translation for 'onlineSavingsMsg'
Learn More

American Biotech Supply Standard Hazardous Location (Explosion Proof) Refrigerator with Natural Refrigerants
GREENER_CHOICE

Catalog No. 22651385
Change view
Click to view available options
Capacity (English):
14 cu. ft.
17 cu. ft.
20 cu. ft.
Energy Usage:
1 to 4 HP
1 to 5 HP
Min. Clearance:
70 in.
75 in.
80 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Energy Usage Min. Clearance
22651385 20 cu. ft. 1 to 4 HP 80 in.
22651383 14 cu. ft. 1 to 5 HP 75 in.
22651384 17 cu. ft. 1 to 4 HP 70 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22651385 Supplier American Biotech Supply Supplier No. ABTHCERP20

Refrigerators are engineered for cold-storage use in environments where volatile conditions could potentially exist. These units meet NFPA and OSHA guidelines Class 1, Division II, Group C & D for storing substances in hazardous locations.

Standard Hazardous Location (Explosion Proof) Refrigerators with Natural Refrigerants are engineered for cold-storage use in environments where volatile conditions could potentially exist. All units include a two year parts and labor warranty. They run on natural, hydrocarbon refrigerant for environmental health and economic efficiency. These units meet NFPA and OSHA guidelines Class 1, Division II, Groups C and D for storing substances in hazardous locations.*


*Explosion Proof Units are designed for environments where external hazards could exists. For storage of volatile substances in normal laboratory conditions, see ABS Flammable Storage Units.

  • Microprocessor Temperature Controller
  • Interior shelving
  • Keyed door lock
  • High density urethane foam cabinet and door insulation
  • Not equipped with line cord and plug; must be hardwired in conduit to 120 VAC power source
  • ETL Listed

Specifications

Type HazLoc Refrigerator
Capacity (English) 20 cu. ft.
Capacity (Metric) 566 L
Temperature Range 1°C to 10°C
Temperature Sensor Microprocessor Temperature Controller
Temperature Stability ±0.1°C
Refrigerant Natural, Hydrocarbon
Door Style Solid Swing
No. of Doors 1
Hinge Side Right
Shelves 5 Adjustable
Cabinet Material Scratch-resistant Painted Steel
Exterior Finish Powder-coated Steel
Insulation Polyurethane Foam
Interior Plastic
Certifications/Compliance cULus
Style Upright
Display Manual
Color White
Condenser Type Fan-forced
For Use With (Application) Cold Storage
Ports None
Warranty Two Year Parts and Labor Warranty
Defrost Cycle
Min. Clearance 80 in.
Monitoring Options Microprocessor Temperature Controller
Compressor Bottom-mounted
Depth (English) Interior 30.75 in.
Depth (Metric) Interior 781 mm
Amperage 3 A
Plug Type NEMA 5-15 P
Voltage 115 V
Energy Usage 1 to 4 HP
Show More Show Less