Learn More
American Biotech Supply Standard Hazardous Location (Explosion Proof) Refrigerator with Natural Refrigerants
Refrigerators are engineered for cold-storage use in environments where volatile conditions could potentially exist. These units meet NFPA and OSHA guidelines Class 1, Division II, Group C & D for storing substances in hazardous locations.
Supplier: American Biotech Supply ABTHCERP17
Description
Standard Hazardous Location (Explosion Proof) Refrigerators with Natural Refrigerants are engineered for cold-storage use in environments where volatile conditions could potentially exist. All units include a two year parts and labor warranty. They run on natural, hydrocarbon refrigerant for environmental health and economic efficiency. These units meet NFPA and OSHA guidelines Class 1, Division II, Groups C and D for storing substances in hazardous locations.*
*Explosion Proof Units are designed for environments where external hazards could exists. For storage of volatile substances in normal laboratory conditions, see ABS Flammable Storage Units.
- Microprocessor Temperature Controller
- Interior shelving
- Keyed door lock
- High density urethane foam cabinet and door insulation
- Not equipped with line cord and plug; must be hardwired in conduit to 120 VAC power source
- ETL Listed
Specifications
| HazLoc Refrigerator | |
| 481 L | |
| Microprocessor Temperature Controller | |
| Natural, Hydrocarbon | |
| 1 | |
| 5 Adjustable | |
| Powder-coated Steel | |
| Plastic | |
| Upright | |
| White | |
| Cold Storage | |
| Two Year Parts and Labor Warranty | |
| 70 in. | |
| Bottom-mounted | |
| 781 mm | |
| NEMA 5-15 P | |
| 1 to 4 HP |
| 17 cu. ft. | |
| 1°C to 10°C | |
| ±0.1°C | |
| Solid Swing | |
| Right | |
| Scratch-resistant Painted Steel | |
| Polyurethane Foam | |
| cULus | |
| Manual | |
| Fan-forced | |
| None | |
| Cycle | |
| Microprocessor Temperature Controller | |
| 30.75 in. | |
| 3 A | |
| 115 V |