missing translation for 'onlineSavingsMsg'
Learn More

Welch™ Standard-Duty Vacuum Pumps

Compact, economical WOB-L™ pumps for aqueous applications

Supplier:  Welch™ 2522B01

 View more versions of this product

Catalog No. 010511A


Only null left

Alert:

May be used with buffer solutions but are not recommended for use with acids, bases or organic vapors.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.


Includes: Liquid trap on inlet, vacuum gauge and adjustment knob, exhaust muffler, suction cup feet, and handles for convenient transportation

Description

Description

Compact, economical WOB-L™ pumps for aqueous applications

  • Ideal for filtration, vacuum ovens, aspiration and chamber roughing
  • Protect against accidental ingestion
  • Wetted parts resist corrosion from moisture

Order Info

For 115V 60Hz. All pumps weigh less than 25 lb. (11.4kg).

Specifications

Chemical Identifiers

CE
115 V 60 Hz
60 Hz
115 V

Specifications

0.12 hp
7.7 kg
CE
2522
5.3 kg
60 Hz
25.4 cm
20.6 cm
100 psi
17 lb.
Liquid trap on inlet, vacuum gauge and adjustment knob, exhaust muffler, suction cup feet and handles for convenient transportation
115 V 60 Hz
11.7 lb.
22 L/min.
10 in.
8.1 in.
8.8 in.
115 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions