Learn More
Brady™ Standard Barricade Tape Roll: Non-Adhesive Lightweight Polyethylene, CAUTION RADIATION HAZARD, Magenta on Yellow
Catalog No. 296357917
Change view
Click to view available options
Color:
Magenta on Yellow
Catalog No. Color
296357917 Magenta on Yellow
1 options
Catalog No. 296357917 Supplier Brady™ Supplier No. 91470
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This durable, lightweight barricade tape is made of non-adhesive, glossy B-912 polyethylene for on-the-spot warnings of potential danger.
  • Non-adhesive Polyethylene (B-912) is a glossy plastic that's ideal for temporary outdoor or permanent indoor use
  • Lightweight tape can be tied, stapled or nailed to posts, fences or metal barricades
  • Visual barricading provides fast on-the-spot warning of potential danger
  • B-912 polyethylene barricade tapes from Brady™ are available in variety of legends to fit your specific needs (each roll/legend is sold separately)
Specifications
Color Magenta on Yellow