Learn More
Stanbio™ QuPID™ Plus hCG Test
Catalog No. SB1230025
Change view
Click to view available options
Quantity:
25 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB1230025 25 Tests
SB1230050 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SB1230025 Supplier Stanbio™ Supplier No. 1230 025KH
Includes
Disposable plastic dropper
Qualitative platform immunoassay device
  • Detects hCG in 3 minutes in urine or 5 minutes in serum
  • Sensitive to 20 mIU/mL or greater
  • Specific monoclonal/polyclonal antibodies ensure early pregnancy detection while preventing cross reactivity with hormones present in patient samples
  • Complete instructions printed directly on each individual test unit's foil pouch
Specifications
Accuracy >99%
Clia Complexity Waived
Detectable Analytes hCG
Sample Type Serum, Urine
Type hCG Pregnancy Test
Includes Complete instructions printed directly on each individual test unit's foil pouch, as well as a disposable Plastic dropper for convenience and ease-of-use
Quantity 25 Tests
Test Time 3 min. for urine, 5 min. for serum