missing translation for 'onlineSavingsMsg'
Learn More

Stanbio™ QuPID™ hCG Test

Catalog No. SB1220050
Change view
Click to view available options
Quantity:
25 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB1220050 50 Tests
SB1220025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SB1220050 Supplier Stanbio™ Supplier No. 1220050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Qualitative platform immunoassay device

  • Dip and read strip format for the visual qualitative detection of hCG in urine
  • Employs a test strip with a unique combination of monoclonal/polyclonal anti-hCG antibody, a colored reagent and two immobilized capture antibodies directed against hCG and mouse IgG respectively
  • Results in 3 minutes
  • Sensitive to 20 mIU/mL hCG in urine
  • Store at room temperature (2–30°C)

Specifications

Accuracy >99%
Clia Complexity Waived
Detectable Analytes hCG
Sample Type Urine
Type hCG Pregnancy Test Kit
Quantity 50 Tests
Test Time 3 min.