Learn More
Stanbio™ hCG Tri-Level Serum Controls
Catalog No. SB1235305
Change view
Click to view available options
Quantity:
5 mL
Catalog No. Quantity
SB1235305 5 mL
1 options
Potential biohazardous material.
Catalog No. SB1235305 Supplier Stanbio™ Supplier No. 1235305
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Potential biohazardous material.
With low-positive, high-positive and negative controls
  • For use with Qu-PID HCG serum tests
Specifications
Type hCG tri-level Serum Controls
Includes High-positive, low-positive and negative controls
Intended Use Qu-PID HCG Serum tests
Quantity 5 mL