missing translation for 'onlineSavingsMsg'
Learn More

StatLab StainSafe Resin Ribbon

Catalog No. 21033458
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21033458 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21033458 Supplier StatLab Supplier No. SSRIBBONZ

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Zebra printer compatible with 75 meters in length

  • 1/2 in. core, 2.5 in. wide
  • 75 meters in length

Specifications

Product Type Printer Ribbon
Color White
For Use With (Equipment) Zebra Label Printer
Material Resin
Print Technology Thermal Transfer
Length (English) 75 m
Width (English) 2.5 in.