Learn More
Sklar Instruments Disposable Stainless Steel Surgical Blades, Sklar #22. Sklar Disposable, Sterile, Stainless Steel Surgical Blade #22, BX100.
Catalog No. 501094395
Encompass
Disposable Stainless Steel Surgical Blades, Sklar, #22. Sklar's Disposable, Sterile, Stainless Steel Surgical Blade #22 is a larger version on the #10 Scalpel Blade. It has a curved cutting edge and a flat, unsharpened back edge. The #22 blade is larger than both the #20 and #21 blades. It is compatible with the #4 series and larger handles. This blade is commonly used in cardiac and thoracic procedures to make incisions in the bronchus. These disposable blades are available in bulk quantities of 100 per box.
Catalog No. 501094395 Supplier Sklar Instruments Supplier No. 063106
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Disposable Stainless Steel Surgical Blades, Sklar, #22. Sklar's Disposable, Sterile, Stainless Steel Surgical Blade #22 is a larger version on the #10 Scalpel Blade. It has a curved cutting edge and a flat, unsharpened back edge. The #22 blade is larger than both the #20 and #21 blades. It is compatible with the #4 series and larger handles. This blade is commonly used in cardiac and thoracic procedures to make incisions in the bronchus. These disposable blades are available in bulk quantities of 100 per box.